Order Entry
United States
Orders LinkContactUsLinkComponent
4-Bromomethyl-6,7-dimethoxycoumarin ≥98.0%

4-Bromomethyl-6,7-dimethoxycoumarin ≥98.0%

4-Bromomethyl-6,7-dimethoxycoumarin ≥98.0%
Supplier:  TCI America
Certificates
4-Bromomethyl-6,7-dimethoxycoumarin ≥98.0%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

[for HPLC Labeling] CAS Number: 88404-25-5 MDL Number: MFCD00011570 Molecular Formula: C12H11BrO4 Molecular Weight: 299.12 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Melting point (°C): 214 Storage Temperature: 0-10°C

  • CAS Number:88404-25-5
  • Formula:C₁₂H₁₁BrO₄
  • MDL Number:MFCD00011570
  • Molecular Weight:299.12 g/mol
  • Melting Pt:214 °C

Documents

Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    98.0% (HPLC)
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $61.00
    $61.00
    Each (100mg)
       $61.00

    Supplier Number: A5570-100MG
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $399.00
    $399.00
    Each (1g)
       $399.00

    Supplier Number: A5570-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.