Order Entry
United States
ContactUsLinkComponent
Anti-Vasopressin V2 Receptor Rabbit Polyclonal Antibody
Anti-Vasopressin V2 Receptor Rabbit Polyclonal Antibody
Catalog # 89365-124
Supplier:  Genetex
Anti-Vasopressin V2 Receptor Rabbit Polyclonal Antibody
Catalog # 89365-124
Supplier:  Genetex
Supplier Number:  GTX70835
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Vasopressin V2 Receptor
  • Antigen Symbol:
    Vasopressin V2 Receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Storage Buffer:
    Phosphate buffered saline, PH 7.7 containing 0.01% sodium azide
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide - KLH conjugated
  • Tested Applications:
    IHC-P
  • Purification:
    Immunogen affinity purified.
  • Cat. No.:
    89365-124
  • Supplier no.:
    GTX70835

Specifications

About this item

The human AVPR2 locus encodes vasopressin receptor type 2, a member of the vasopressin/oxytocin family subfamily. V2 receptor has been shown to concentrate the urine and maintain water homeostasis by responding to the pituitary hormone arginine vasopressin (AVP). Loss of gene function results in the disease Nephrogenic Diabetes Insipidus (NDI). When extrarenal V2 receptors are stimulated by infusion of a V2 selective agonist (dDAVP), a variety of clotting factors are released into the bloodstream. The physiologic importance of this property is not known; its absence does not appear to be detrimental in NDI patients. The V2 receptor is expressed in the kidney tubule, predominantly in the distal convoluted tubule and collecting ducts. The V2 Receptor is also expressed outside the kidney although its tissue localization is uncertain. AVPR2 expression has also been described in brain, breast, fetal lung tissue and lung cancer associated with alternative splicing. ESTs have been isolated from normal lung and pancreatic cancer libraries.

Type: Primary
Antigen: Vasopressin V2 Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human