Anti-TIMP2 Mouse Monoclonal Antibody [clone: T2-101]
89367-062
: Genetex
- Antibody Type:Primary
- Antigen Name:Tissue inhibitor of metalloproteinases 2
- Antigen Symbol:TIMP2
- Clonality:Monoclonal
- Clone:T2-101
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:500 μl
- Gene ID:7077
- Antigen Synonyms:tissue inhibitor of metalloproteinases 2|metalloproteinase inhibitor 2|TIMP-2|tissue inhibitor of metalloproteinase 2
- Storage Buffer:10mM PBS, pH7.4, 0.2%BSA, 0.09% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:Native human pro-MMP-2/TIMP-2 complex purified from A2058 culture medium.
- Tested Applications:IP, WB
- Purification:Protein G purified
- Cat. No.:89367-062
- Supplier no.:GTX23161
Mouse monoclonal [T2 - 101] to TIMP2
Immunoprecipitation: use at a concentration of 2?g / mg. Western Blotting: use at a concentration of 1 - 2?g/ml for 2hrs at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of 21 kDa, which corresponds to the predicted molecular weight of TIMP-2.
Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human