Anti-SYN Rabbit Polyclonal Antibody
Catalog # 89366-734
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Syntrophin
- Antigen Symbol:SNTA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:281510
- Antigen Synonyms:MGC111389|59-DAP |DAPA1B |TIP-43|A1B |Syntrophin-2 |FLJ22442|BSYN2|SNT2|SNT2B1
- Modification:phospho-Ser62,67
- Storage Buffer:100 µl in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 µg per ml BSA and 50% glycerol.
- Molecular Weight:78 kDa
- Storage Temperature:For long-term storage, aliquot and store at -20 C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser62/67 conjugated to KLH
- Tested Applications:WB
- Purification:Antigen affinity purification
- Cat. No.:89366-734
- Supplier no.:GTX82591
Specifications
About this item
Rabbit polyclonal antibody to Synapsin (phospho Ser62,67)
Type: Primary
Antigen: SYN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat