Order Entry
United States
ContactUsLinkComponent
Anti-SUCLG1 Rabbit Polyclonal Antibody
Anti-SUCLG1 Rabbit Polyclonal Antibody
Catalog # 89355-808
Supplier:  Genetex
Anti-SUCLG1 Rabbit Polyclonal Antibody
Catalog # 89355-808
Supplier:  Genetex
Supplier Number:  GTX109215

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Succinate-CoA Ligase, GDP-forming, Alpha Subunit
  • Antigen Symbol:
    SUCLG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    8802
  • Antigen Synonyms:
    SUCLA1|GALPHA|MTDPS9
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    36 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Full length human SUCLG1 Recombinant protein.
  • Tested Applications:
    ICC/IF, IHC-P, WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89355-808
  • Supplier no.:
    GTX109215

Specifications

About this item

Rabbit Polyclonal antibody to SUCLG1 (succinate-CoA ligase, alpha subunit)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SUCLG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human