Order Entry
United States
ContactUsLinkComponent
Anti-PDE4C Rabbit Polyclonal Antibody
Anti-PDE4C Rabbit Polyclonal Antibody
Catalog # 89367-344
Supplier:  Genetex
Anti-PDE4C Rabbit Polyclonal Antibody
Catalog # 89367-344
Supplier:  Genetex
Supplier Number:  GTX14608

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Phosphodiesterase 4C
  • Antigen Symbol:
    PDE4C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    5143
  • Antigen Synonyms:
    DPDE1
  • Storage Buffer:
    Preservative: 0.02% Thimerosal / Merthiolate
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Synthetic cyclic peptide (carboxy-terminal peptide common to all PDE4C proteins).
  • Tested Applications:
    IP, WB
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    89367-344
  • Supplier no.:
    GTX14608

Specifications

About this item

Cyclic nucleotides are hydrolyzed and compartmentalized by a family of enzymes called phosphodieterases. cAMP-specific phosphodiesterase type-4 (PDE4) family is comprised of 4 genes (PDE4 A, B, C and D) with multiple splice variants generated by RNA splicing and use of alternate initiation sites. The cAMP-dependent protein kinase A modulates the enzyme activity of some PDE4 family members by phosphorylation/dephosphorylation mechanisms. Such short-term regulatory mechanisms are necessary for rapid changes in the cAMP levels in cells.

IP: Use at a dilution of 1/20. WB: Use at a dilution of 1/500. Predicted molecular weight: 80 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PDE4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human