Anti-IRF9 Rabbit Polyclonal Antibody
89366-310
: Genetex
- Antibody Type:Primary
- Antigen Name:IRF9
- Antigen Symbol:IRF9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:10379
- Antigen Synonyms:IRF-9|p48|ISGF3|ISGF3G
- Storage Buffer:PBS with 2% sucrose
- Storage Temperature:Store at -20 °C or below. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human ISGF3G
- Tested Applications:WB
- Purification:Rabbit Ig G is purified by peptide affinity chromatography method.
- Cat. No.:89366-310
- Supplier no.:GTX77807
Rabbit Polyclonal Antibody to ISGF3G
WB: Suggested dilution at 0.005-0.05 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.
Type: Primary
Antigen: IRF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human