Order Entry
United States
ContactUsLinkComponent
Anti-IL12 Mouse Monoclonal Antibody [clone: 1-1A4]
Anti-IL12 Mouse Monoclonal Antibody [clone: 1-1A4]
Catalog # 89360-760
Supplier:  Genetex
Anti-IL12 Mouse Monoclonal Antibody [clone: 1-1A4]
Catalog # 89360-760
Supplier:  Genetex
Supplier Number:  GTX20050
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Interleukin 12
  • Antigen Symbol:
    IL12
  • Clonality:
    Monoclonal
  • Clone:
    1-1A4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 μg
  • Gene ID:
    3593
  • Antigen Synonyms:
    IL-12|cytotoxic lymphocyte maturation factor 35 kDa subunit|NK cell stimulatory factor chain 1|interleukin-12 subunit alpha|p35|natural killer cell stimulatory factor 1|interleukin-12 alpha chain|interleukin 12|IL-12 subunit p35|CLMF p35|cytotoxic lymphocyte maturation factor 1|subunit p35|35 kD subunit|NF cell stimulatory factor chain 1
  • Storage Buffer:
    Phosphate-buffered saline, pH 7.2, no preservative added
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.7 mg/ml
  • Immunogen:
    Recombinant human interleukin-12 produced in Baculovirus.
  • Tested Applications:
    Radio Immunoassay
  • Purification:
    Protein A affinity purified
  • Cat. No.:
    89360-760
  • Supplier no.:
    GTX20050

Specifications

About this item

IL12 is a heterodimeric cytokine comprised of p35 and p40 subunits. IL12 plays a central role in mediating cell mediated immunity, promoting differentiation of CD4+ T cells to the Th1 subset and of CD8+ T cells into mature cytotoxic T lymphocytes (CTLs). IL12 is a potent stimulator of NK cells. IL12 therapy has been suggested as a method of enhancing cytocidal anti tumour immune responses.

For ELISA: Use at an assay dependent concentration. For RIA: Use at an assay dependent concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL12
Clonality: Monoclonal
Clone: 1-1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human