Order Entry
United States
ContactUsLinkComponent
Anti-DDR2 Rabbit Polyclonal Antibody
Anti-DDR2 Rabbit Polyclonal Antibody
Catalog # 89349-678
Supplier:  Genetex
undefined
Anti-DDR2 Rabbit Polyclonal Antibody
Catalog # 89349-678
Supplier:  Genetex
Supplier Number:  GTX102651

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Discoidin Domain receptor tyrosine kinase 2
  • Antigen Symbol:
    DDR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    4921
  • Antigen Synonyms:
    Receptor protein-tyrosine kinase TKT|receptor-related 3|Neurotrophic tyrosine kinase|NTRKR3|DDR2|Discoidin domain receptor 2|TKT|Tyrosine-protein kinase TYRO 10
  • Storage Buffer:
    1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    97 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the C-terminus region of human DDR2. The exact sequence is proprietary.
  • Tested Applications:
    IHC-P
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89349-678
  • Supplier no.:
    GTX102651

Specifications

About this item

Rabbit Polyclonal antibody to DDR2 (discoidin domain receptor tyrosine kinase 2)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DDR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human