Anti-TKFC Rabbit Polyclonal Antibody
100 μl
Catalog # 89349-854
Supplier: Genetex
Supplier #: GTX103102
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit Polyclonal antibody to DAK (bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TKFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:100 μl
- Clonality:Polyclonal
- Molecular Weight:59 kDa
- Purification:Purified by antigen-affinity chromatography.
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- ImmunoFluorescence:Yes
- Tested Applications:ICC/IF, IHC-P, WB
- Isotype:IgG
- Host:Rabbit
- Concentration:0.76mg/ml
- Cat. No.:89349-854
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:TKFC
- Gene ID:26007
- Immunogen:Recombinant protein encompassing a sequence within the center region of human DAK. The exact sequence is proprietary.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:NET45|DAK
- Antigen Name:Triokinase And FMN Cyclase
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



