To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD81 molecule
- Antigen Symbol:CD81
- Clonality:Monoclonal
- Clone:1D6
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Sheep,Goat,Chimpanzee,Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:975
- Antigen Synonyms:CD81 antigen|S5.7|TSPAN28|26 kDa cell surface protein TAPA-1|CVID6|Tetraspanin-28|TAPA1|Target of Antiproliferative 1|TSPAN-28
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage Temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Aggregated OCI-LY8 cells (Human).
- Tested Applications:Fluorescence-Activated Cell Sorting
- Purification:Purified IgG prepared by affinity chromatography on Protein A from tissue culture supernatant
- Cat. No.:89363-528
- Supplier no.:GTX43505
Specifications
About this item
Mouse monoclonal antibody [1D6] to CD81
Recommended Starting Dilutions: Flow Cytometry: 1/5 - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: