- Antibody Type:Primary
- Antigen Name:Ghrelin
- Antigen Symbol:GHRL
- Clonality:Monoclonal
- Clone:Ghr5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:51738
- Antigen Synonyms:ghrelin 28|obestatin|ghrelin 27|Growth hormone-releasing peptide|MTLRP|Protein M46|Motilin-related peptide|Appetite-regulating hormone
- Storage Buffer:Phosphate-buffered saline, pH 7.4, containing 0.1% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:2 mg/ml
- Immunogen:Synthetic peptide (Human)
- Tested Applications:ELISA, WB
- Purification:Protein G purified
- Cat. No.:89350-890
- Supplier no.:GTX13983
Ghrelin is an endogenous ligand for the growth hormone secretagogue receptor and is involved in regulating growth hormone release from the pituitary. It has an appetite stimulating effect, induces adiposity, and stimulates gastric acid secretion, and it is involved in growth regulation. Ghrelin is synthesized as a preprohormone, then proteolytically processed to yield a 28 amino acid peptide. An interesting and unique modification is imposed on the hormone during synthesis in the form of an n octanoic acid bound to one of its amino acids. This modification is necessary for biologic activity.
ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 13 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GHRL
Clonality: Monoclonal
Clone: Ghr5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human