To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Calpain 2
- Antigen Symbol:CAPN2
- Clonality:Monoclonal
- Clone:1E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Bovine
- Western Blot:Yes
- Size:50 μg
- Gene ID:824
- Antigen Synonyms:CANP2|CANPL2|mCANP|FLJ39928|Millimolar-calpain|EC 3.4.22.53|M-calpain|CANPml
- Storage Buffer:Preservative: 0.02% Sodium Azide; Constituents: PBS
- Storage Temperature:Upon receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:2 mg/ml
- Immunogen:Bovine cardiac Calpain 2.
- Tested Applications:ELISA, ICC/IF, WB
- Purification:Immunogen affinity purified
- Cat. No.:89360-886
- Supplier no.:GTX20713
Specifications
About this item
Proteases influence intracellular signaling pathways and regulate cytoskeleton organization. It was previously described that novel autoantibodies to calpastatin (endogenous inhibitor for calpain) were detected in patients with rheumatoid arthritis (RA) and other disorders. Since calpain is thought to mediate inflammatory process and cartilage destruction a screening of anti-calpain antibodies in human patient sera is recommended.
Western blot: 1 ?g/ml. Immunofluorescence: use the antibody after 2% paraformaldehyde fixation in a concentration of 0.5 ?g/ml.
Type: Primary
Antigen: CAPN2
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine