Order Entry
United States
ContactUsLinkComponent
Anti-MDM2 Rabbit Polyclonal Antibody
Anti-MDM2 Rabbit Polyclonal Antibody
Catalog # 89348-522
Supplier:  Genetex
undefined
Anti-MDM2 Rabbit Polyclonal Antibody
Catalog # 89348-522
Supplier:  Genetex
Supplier Number:  GTX100531

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Mouse Double Minute 2
  • Antigen Symbol:
    MDM2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • ChIP:
    Yes
  • Gene ID:
    4193
  • Antigen Synonyms:
    1700007J15Rik|Double minute 2 protein|AA415488|HDMX|Ubiquitin-protein ligase E3 Mdm2|Mdm-2|ACTFS|Oncoprotein Mdm2|p53-binding protein Mdm2
  • Storage Buffer:
    1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    56 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.74 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human MDM2. The exact sequence is proprietary.
  • Tested Applications:
    ICC/IF, IHC-P, WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89348-522
  • Supplier no.:
    GTX100531

Specifications

About this item

Rabbit Polyclonal antibody to MDM2 (Mdm2 p53 binding protein homolog)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MDM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human