Order Entry
United States
ContactUsLinkComponent
ReadyPrep Protein Extraction Kit (Membrane II), Bio-Rad
ReadyPrep Protein Extraction Kit (Membrane II), Bio-Rad
Catalog # 101644-016
Supplier:  Bio-Rad
CAS Number:  
undefined
ReadyPrep Protein Extraction Kit (Membrane II), Bio-Rad
Catalog # 101644-016
Supplier:  Bio-Rad
Supplier Number:  1632084
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ReadyPrep Protein Extraction Kit (Membrane II)
  • Size:
    10 preps
  • Cat. No.:
    101644-016
  • Application:
    For Complex Membrane Proteins, Na Carbonate Extraction
  • Supplier no.:
    1632084

Specifications

About this item

The ReadyPrep protein extraction kit (membrane II) applies sodium carbonate extraction methods (Fujiki et al

1982, Molloy et al. 2000) to fractionate and isolate membrane proteins. The ReadyPrep protein extraction kit (membrane II) isolates complex proteins that contain more than two transmembrane domains or those that are resistant to extraction by the ReadyPrep protein extraction (membrane I) kit. Requires ultracentrifugation. Includes a strongly chaotropic, IEF-compatible rehydration/sample buffer that contains the detergent ASB-14 to efficiently solubilize proteins. References: Fujiki Y et al. -1982. Isolation of intracellular membranes by means of sodium carbonate treatment: application to endoplasmic reticulum. J Cell Biology 93, 97–102. Molloy MP et al. -2000. Proteomic analysis of the Escherichia coli outer membrane. Eur J Biochem 267, 2871–2881.

Ordering information: Includes strong detergent ASB-14.