Orders LinkContactUsLinkComponent
VWR® Vortex Mixers | Avantor
VWR® Vortex Mixers
VWR® Vortex Mixers are commonly used in laboratories to mix small liquid volumes for cell disruption or homogenization. Analog or digital units precisely control the operational speed and time to achieve reproducible results. Capable of mixing single or multiple samples simultaneous, VWR vortex mixers accommodate test tubes, vials, centrifuge tubes, flasks and even microplates. Constructed of rugged, corrosion-resistant materials, our units will withstand heavy everyday usage.
  • Mini Vortexers are great for simple mixing.
  • Heavy-Duty and Microplate Vortex mixers are the ideal solution for powerful, continuous duty vortexing. These mixers have a unique locking design for the accessories allowing for secure high-speed mixing.
  • The Multi-Tube Vortexers offer powerful, hands-free mixing for up to 50 samples at one time.
Product selection