Orders LinkContactUsLinkComponent
Dry-Block-Heater540-390.jpg
VWR® Dry Block Heaters
VWR® Dry Block Heaters are compact, versatile, multi-purpose units available in a variety of block capacities with choices of digital and analog controls. All models offer exceptional temperature stability and uniformity. Digital models provide repeatable results and feature a temperature calibration mode. Ideal for incubation and activation of cultures, enzyme reactions, immunoassays and a variety of other laboratory procedures.
VWR® Dry Block Heaters | Avantor
Each model accepts interchangeable modular blocks with over 40 options from which to choose. Blocks feature a close tube and block contact for maximum heat transfer and a thermometer well for measuring block temperature.
AdvancedDryBlockHeaters400-495.jpg
VWR® Advanced Dry Block Heaters
Our economical, versatile and compact block heaters are designed for applications that require repeatable results and superior temperature stability.
HeatedLid400-495.jpg
VWR® Advanced Dry Block Heater with Heated Lid
Provides exceptional temperature uniformity and stability while reducing condensation on sample lids.
Standard-Dry-Block-Heaters400-495.jpg
VWR® Standard Dry Block Heaters
Multipurpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays and a variety of other laboratory procedures.
BlockHeatersAndBlocks4jpg.jpg
Modular blocks are constructed of anodized aluminum. The close contact of tube-block-walls allows for maximum heat retention.
Contact us today to discuss your needs. Our team is ready to help.