Orders LinkContactUsLinkComponent
20647532-f.jpg
Decontaminant and Detergent Products
Surgical Site Infections (SSIs) are an increasingly common issue for hospitalized patients, causing over one-third of all hospital-acquired infections in surgical patients, according to the NIH.
Since every stage of the cleaning process requires critical and expert attention, Sklar provides a full line of care and cleaning agents to decontaminate, sterilize, lubricate, and protect your surgical instruments and equipment against pathogens like H1N1, HIV-1, HSV, VRE, Rotaviruses, Hep B&C as well as TB, MRSA, and A. Baumannii.
All cleaning agents are suitable for use on metals, glassware, plastic, and rubber. SDS sheets and certificates of sterility are also provided.
Product selection
sklar_decon_90x90.jpg
Safely sterilize soiled instruments in 2 minutes with an industry-leading kill time against pathogens. Available in gallons, sprays, and wipes.
sklar_zyme_90x90.jpg
Remove heavy organic debris build-up from cloth, steel, plastic, and rubber with fast acting, biodegradable, non-corrosive cleaners.
sklar_lube_90x90.jpg
Extend the life and functionality of your instruments with easy-to-use lubricants that inhibit rust and corrosion, even in hard-to-reach areas.
sklar_polish_90x90.jpg
Liquid and powder polishes that are non-abrasive, non-corrosive, and work with manual, automatic, and ultrasonic cleaning methods.