Orders LinkContactUsLinkComponent
Labconco® - Chemical Fume Hoods | Avantor
Labconco's fume hoods maximize containment to keep hazardous fumes away from you and your lab all while maintaining efficient operation. Efficient operation means less air exhausted from the lab, which translates to saving energy.
Labconco's Protector XStream® and Premier® laboratory fume hoods are made with high quality components and precision manufacturing to ensure long lasting performance. Worksurfaces, stands and accessories to meet your specific needs are available. Made in the USA. ETL certified meeting UL Standard 61010-1, UL-1805 and CAN/CSA C22.2 No 61010.1. See how a Labconco fume hood can offer high performance protection while lowering your energy usage.