Orders LinkContactUsLinkComponent
Hero1_sf_2222.jpg
PUREZERO* Cleanroom Gloves
Your gloves have a big job to do, protecting your workers as well as your product and your process. Plus you need a reliable supply to avoid operations disruption. That’s why we created HALYARD* PUREZERO* Cleanroom Gloves.
PUREZERO* Cleanroom Gloves are ideal for applications that involve handling delicate equipment in microelectronics, semiconductors, optics, pharmaceutical and medical device manufacturing applications.   These gloves are designed to exact standards, to help you manage the risks associated with user comfort and protection, product contamination and supply chain resiliency.
PUREZERO* Cleanroom Gloves | Avantor
Block1_sf_2222.jpg
HALYARD* PUREZERO* HG3 WHITE SGX* Nitrile Cleanroom Gloves
PUREZERO* HG3 Smooth White Nitrile Cleanroom Gloves are available in non-sterile.  They measure 12 inches long with a smooth finish and feature a beaded cuff to aid in donning and help prevent roll down.  The maximum particle count is >0.5μm / cm2 <950 IEST RP-CC005.
Block2_sf_2222.jpg
HALYARD* PUREZERO* HG3 Light Blue Non-Sterile Nitrile Gloves
Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms environments. PUREZERO* HG3 Light Blue Nitrile Cleanroom Gloves are non-sterile and 12 inches long with a tacky grip and feature a beaded cuff to aid in donning and help prevent roll down.  The max particle count is >0.5μm / cm2 <1200 (IEST RP-CC005).
Block3_sf_2222.jpg
HALYARD* PUREZERO* HG3 White Non-Sterile Nitrile Gloves
Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms environments. PUREZERO* HG3 White Nitrile Cleanroom Gloves are non-sterile and 12 inches long with a tacky grip and feature a beaded cuff to aid in donning and help prevent roll down.  The max particle count is >0.5μm / cm2 <950 (IEST RP-CC005).
Block4_sf_2222.jpg
HALYARD* PUREZERO* HG3 Light Blue Sterile Nitrile Gloves
Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms environments. PUREZERO* HG3 Light Blue Nitrile Cleanroom Gloves are sterile and 12 inches long with a smooth grip and feature a beaded cuff to aid in donning and help prevent roll down. The max particle count is >0.5μm / cm2 <1200 (IEST RP-CC005).
Block5_sf_2222.jpg
HALYARD* PUREZERO* HG3 White Sterile Nitrile Gloves
Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms environments. PUREZERO* HG3 White Nitrile Cleanroom Gloves are sterile and 12 inches long with a smooth grip and feature a beaded cuff to aid in donning