Avantor® Seradigm Sera
Cell culture experts everywhere are discovering how well their cells thrive on nutritionally superior sera from Avantor Seradigm. Seradigm’s collection and manufacturing techniques preserve more of the naturally-occurring growth factors in sera that cells require to thrive. Our proven track record for performance and consistency enables sera users to switch to a brand that yields more of the results they seek – no matter the application, scale, or budget.
Cells Choose Seradigm
The collection and manufacturing techniques used for our sera preserve more of the naturally-occurring growth factors that cells require to thrive. Our proven track record for performance and consistency enables sera users to switch to a brand that yields more of the results they seek – no matter the application, scale, or budget.
- Nutritionally superior sera
- Proven performance and consistency
- Supply stability with industry-leading traceability and quality
- Unmatched operational support and service
Sera Product Differentiation Table
Find the right sera for your application according to country of origin, product features and specifications.
Avantor offers a trusted supply chain of sera with…
Quality & Choice
- Proven sera performance & consistency
- Manufacturing methods result in lot-to-lot and bottle-to-bottle consistency
- Quality you can see in the crystal clear, vibrant color of each bottle of sera
Expertise
- Nutritionally superior sera
- Collection & manufacturing techniques preserve more naturally-occurring growth factors found in serum
- Industry-leading traceability & quality
- Independent labs used for all lot release criteria
- ISIA traceability certification confirms truth in labeling
Convenience
- Strong supply partnerships and raw material supply chain provide long-term assurance of supply
Customization
- Unmatched operational sera support & service
- Flexibility in meeting each customer’s unique needs
- Multiple lot sampling, large volume reserves, and storage solutions
- Proven track record of operational excellence, support, and on-time delivery
Have questions about Avantor Seradigm?
Here is everything you need to know, from comparing Seradigm products, literature, how to request a sample, as well as detailed information on origin and traceability.
View the entire line of Avantor Seradigm FBS products and compare country of origin, product features, and specifications.
Need more information, have a question, or would like a product sample sent to you? We’re here to help.
Literature
Download and read product literature on the entire Avantor Seradigm FBS product line.
The Oritain verification of product origin provides enhanced traceability and trust for Avantor Seradigm Fetal Bovine Serum (FBS).
Access Avantor’s ISIA Certification Statement as well as Seradigm Product Information and Material Safety Data Sheets.
Contact Us
Improve your serum experience with Avantor Seradigm. Contact us today to get answers to your questions or receive a sample.
Product selection
Australia Origin FBS combines outstanding performance and quality with ease of regulatory access globally, suitable for a variety of applications.
Ultimate Grade FBS is the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology.
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a variety of applications.
Ultra Low IgG FBS meets a specification of ≤5μg/mL for applications where naturally occurring levels of IgG are too high.
Select Grade FBS is an affordable, quality product providing consistent performance for cost-sensitive applications.
Related content
Ready to move science forward?
We're here to help.