Mobile Phase Preparation
These Solvents and Reagents have been custom designed to meet the high requirements of PFAS Analysis, and are suitable for use for different substances.
Products
You need a solvent supplier you can trust. All our solvents are tested for suitability for HPLC Analysis.
All LC-MS Solvents are tested with Reserpine to ensure very low impurity signals in high-sensitivity spectra to maintain integrity.
Wide range of Solvents enginereed specifically for Pesticide residue analysis by Gaz Chromatography.