EPA Method 537.1 for Drinking water
Determination of selected PFAS in Drinking water by solid phase extraction and Liquid Chromatography/tandem mass spectrometry (LC-MS/MS).
Products
PFAS friendy vials
Specific polypropylene vials recommended when analysing polar compounds /analytes and dimensionally verified/inspected by automated optical and gauges to ensure critical dimensions and tolerances.
Caps for PFAS analysis
SPE for PFAS analysis
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.