Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4 Components for a Successful PCR Setup | Avantor
4 Components for a Successful PCR Setup
Polymerase Chain Reaction (PCR) is a laboratory technique that lets scientists amplify specific DNA sequences from a small amount of starting material. The success of PCR depends on several factors, including the DNA template’s quality, the choice of primers, the reaction conditions, and the setup’s accuracy.
This article will discuss four essential components that help ensure a successful PCR setup, from selecting the right reagents and equipment to optimizing reaction conditions. Following these steps can increase the chances of obtaining reliable and reproducible PCR results for your research or diagnostic applications.
What is a standard PCR protocol?
A standard PCR protocol is a set of guidelines for performing polymerase chain reaction experiments. The protocol includes the standard polymerase chain reaction setup involving several critical steps for success. Here's a brief overview of the steps involved in a standard PCR setup:
  • Gather Materials
The first step is to gather all the necessary materials and equipment for the PCR reaction. It includes DNA templates, primers, Taq polymerase, deoxynucleotide triphosphates (dNTPs), buffer and water.
  • Prepare the Reaction Mix
The reaction mix is prepared by mixing the components correctly. The concentration of each component in the reaction mix is critical to the PCR’s success. The reaction mixture is then aliquoted into PCR tubes or plates.
  • Add Template DNA
The DNA template is added to the reaction mix and can vary depending on the application. It typically ranges from 1-100 ng.
  • Perform Thermal Cycling
The PCR reaction is conducted in a thermal cycler that provides a series of temperature-controlled cycles. Each cycle consists of three steps: denaturation, annealing and extension.
The denaturation step involves heating the reaction mix to a high temperature (usually 94-98°C) to separate the double-stranded DNA into single strands. The annealing step involves cooling the reaction mix to a temperature specific to the primers to let them bind to their complementary DNA sequences.
The extension step involves heating the reaction mix to a temperature optimal for the Taq polymerase to extend the primers and synthesize new DNA strands.
  • Analyze the PCR Products
The final step involves analyzing the PCR products to conduct the amplification of the target DNA sequence. You can do it by agarose gel electrophoresis, sequencing, or other methods.
Essential PCR Components
Polymerase Chain Reaction (PCR) is a molecular biology process amplifying a specific segment of DNA. The components of PCR include:
Template DNA: The DNA sample that requires amplification.
Primers: These are short DNA sequences designed to match specific regions on either side of the DNA segment. They serve as starting points for DNA replication.
Taq Polymerase: These enzymes are responsible for copying the DNA template and can withstand high temperatures during PCR.
Deoxynucleotides (dNTPs): dNTPs are building blocks used by Taq polymerase to synthesize new DNA strands and generally include Adenine, Guanine, Cytosine, and Thymine.
Buffer Solution: The solution provides the optimal chemical environment for Taq polymerase to work efficiently.
Thermal Cycler: A laboratory instrument that cycles through different temperatures to create the optimal conditions for PCR amplification.
Together, these essential components allow for the precise and reliable amplification of specific DNA sequences.
PCR template
In the Polymerase Chain Reaction (PCR), the DNA template is the starting material for amplifying a specific DNA sequence. The PCR template DNA provides the sequence information for the PCR primers to anneal and initiate amplification by the DNA polymerase. The importance of the DNA template in PCR cannot be overstated, as it is the basis of specificity and accuracy in the entire PCR process.
The DNA template can be genomic DNA, cDNA, plasmid DNA, or any other type of DNA that contains the target sequence of interest. The quality and purity of the template DNA are critical factors that can impact the success of the PCR reaction.
DNA quality is important because it determines the ability of primers to anneal and the efficiency of the polymerase to amplify the target sequence. A high-quality DNA template with minimal impurities will improve PCR efficiency and reproducibility.
The DNA template is also important in determining the specificity of the PCR reaction. Specificity refers to the ability of the PCR reaction to amplify only the desired target sequence without any non-specific amplification. The specificity of the PCR reaction depends on the PCR primers' design, which is specific to the target sequence.
However, the quality of the DNA template is also critical in ensuring specificity. A low-quality DNA template may contain impurities or degradation products that can interfere with the specificity of the PCR reaction, resulting in non-specific amplification.
In addition, the quantity of the DNA template is also an important factor to consider in PCR. The quantity of the template DNA must be optimized to ensure enough starting material for efficient amplification while avoiding the risk of inhibition of the PCR reaction due to excess template DNA.
PCR Primer
PCR primer, also known as primer DNA PCR, is a short strand of DNA that serves as a starting point for DNA synthesis in the polymerase chain reaction (PCR). The importance of the PCR primer lies in its ability to accurately and specifically amplify the DNA sequence of interest in PCR protocols.
PCR primer design is critical for the success of PCR, as the primer must be complementary to the template DNA sequence at the desired amplification site. The primer should also have a melting temperature (Tm) appropriate for the PCR reaction conditions and avoid self-complementarity or complementarity with other primers in the reaction mixture.
During PCR, the primer anneals to the single-stranded DNA template, providing a site for the DNA polymerase to synthesize the complementary strand. The DNA polymerase extends the primer by adding nucleotides in the 5' to 3' direction, creating a new strand of DNA complementary to the template strand.
PCR primers are critical for PCR protocols, as they determine the specificity and sensitivity of the amplification reaction. Specificity is essential to ensure that only the target DNA sequence is amplified. At the same time, sensitivity is important to ensure that the amplification reaction is efficient and detects even low amounts of target DNA.
In summary, the PCR primer is a critical component of PCR protocols that determines DNA amplification's accuracy, sensitivity, and specificity. Careful design and selection of PCR primers are essential for the success of PCR experiments in research, clinical, and diagnostic applications.
DNA Polymerase
Polymerase Chain Reaction (PCR) is a powerful molecular biology technique to amplify a specific DNA sequence. The technique relies on the activity of an enzyme called DNA polymerase, which is a crucial component of the PCR reaction.
The primary role of DNA polymerase in PCR is to synthesize new strands of DNA by adding nucleotides to a pre-existing template strand. During the PCR process, the DNA polymerase is typically a thermostable enzyme, which means it can withstand the high temperatures required for the reaction.
The most commonly used DNA polymerase in PCR is Taq polymerase, derived from the bacterium Thermus aquaticus, which has a high processivity and fidelity.
In PCR, the DNA polymerase is responsible for synthesizing new strands of DNA during each cycle of the reaction. The process starts with the denaturation of the DNA template at a high temperature, separating the two double-helix strands.
After the template strands are separated, the primers anneal to the complementary regions of the template DNA, and the DNA polymerase extends the primers to synthesize new strands of DNA. This process is repeated through multiple cycles, resulting in a geometric amplification of the DNA target.
The importance of DNA polymerase in PCR cannot be exaggerated.. The PCR process would be impossible without a highly efficient and specific DNA polymerase. DNA polymerase is responsible for the accuracy and efficiency of PCR, which are key factors in the success of the technique.
The fidelity of the DNA polymerase helps ensure that the amplified DNA is a replica of the original template. At the same time, the high processivity of the enzyme allows for the rapid and efficient synthesis of new strands of DNA.
Taq DNA polymerase is a highly thermostable enzyme derived from the bacterium Thermus aquaticus and is widely recognized as the best-known enzyme used for Polymerase Chain Reaction (PCR). There are several reasons why Taq DNA polymerase is so commonly used in PCR:
  • High Processivity
Taq polymerase has a high processivity, meaning it can add nucleotides to the growing DNA strand without falling off the template. This results in more efficient DNA synthesis, which is important for the success of the PCR reaction.
  • High Thermostability
Taq polymerase is highly thermostable, which means it can withstand high temperatures during the PCR reaction. The enzyme is active at 72-74°C, the optimal temperature for extending PCR primers.
  • Lack of 3’ to 5’ Exonuclease Activity
Taq polymerase lacks the 3' to 5' exonuclease activity, so it cannot proofread and correct errors during DNA synthesis. However, this feature is often beneficial for PCR amplifying complex targets or templates with high GC content, which can be prone to secondary structure formation.
  • Reliability
Taq polymerase is a well-established and widely used enzyme in PCR, with a long history of successful applications in various research fields. It makes it a highly reliable and trusted enzyme for PCR applications.
  • Availability
Taq polymerase is widely available from various commercial vendors, making it accessible to researchers worldwide.
Considerations for a successful PCR Setup
A successful PCR setup can help obtain successful results. Here are some considerations to keep in mind for a successful PCR setup:
  • Template DNA: It’s important to ensure the quality and quantity of the template DNA used for PCR. A good starting point is to use high-quality DNA, free from inhibitors and at an appropriate concentration.
  • Primer Design:  Another consideration is using appropriate primers specific to the target sequence, having similar melting temperatures (Tm), and being free from self-complementarity or hairpin loops.
  • PCR Buffer and Enzyme: The third factor or consideration is using a PCR buffer and enzyme compatible with the primers and template DNA. You can also use high-quality PCR reagents from reputable suppliers.
  • MgCl2 Concentration: Optimize the MgCl2 concentration in the PCR reaction. The concentration of MgCl2 should be optimized to enhance the binding of the primers to the template DNA and improve the PCR fidelity.
  • Annealing Temperature: The PCR reaction’s annealing temperature is another important consideration that requires optimization to ensure specific binding of the primers to the template DNA.
  • PCR Cycling Conditions: Optimize the PCR cycling conditions, including denaturation, annealing, extension times, and temperatures. Additionally, you must ensure that the PCR cycling conditions are appropriate for the primers and template DNA.
  • Reaction Volume: Ensure the correct reaction volume is used for the PCR. The volume of the PCR reaction should be optimized to ensure efficient amplification of the target sequence.
  • Sample Preparation: Carefully prepare the samples for PCR, including DNA extraction, purification, and quantification. Avoid contamination of the samples with extraneous DNA or other contaminants.
For a successful PCR setup, partner with Avantor Sciences. Our expert team recommends using high-quality samples, optimizing reagent handling, primer and probe concentrations, and ensuring accurate thermocycler settings.
Don't forget to use positive and negative controls to validate your results and follow good laboratory practices to minimize contamination risks. Let us help you achieve reliable and reproducible PCR results every time. Contact us today to learn more.
Sources:
1. National Library of Medicine - Research Techniques Made Simple: Polymerase Chain Reaction (PCR)
2. National Library of Medicine - Polymerase Chain Reaction