DILC I-100(Direct Inject LC Instrument Module Includes 0.6 mL mixing chamber, 1 uL sample loop, 0.2 um pre-column filter, 210 x 0.12 mm capillary tubing) 1 * 1 items
Catalog # MSPP1001-1003
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together