SPECIFICATION INCLUDES THE BELOW: 1 x STV22-0109 6T207 bench construction fume cupboard width [mm] 2100 depth [mm] 900 working height [mm] 900 height [mm] 2700 bottom part 1 x supporting construction for fume cupboard - 2100mm wide as H- frame with s 1 * 1 items
Catalog # WALDOF-Q5961
Supplier: WALDNER LABOREINRICHTUNG