Order Entry
United Kingdom
ContactUsLinkComponent
Recombinant Canine parvovirus type 2 Capsid protein VP2 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. H ost: E. coli. Endotoxin Level: Not Tested. Species: Canine parvovirus type 2 (CPV-2). Target Name: Canine parvovirus 1 * 100 µG
  MSPPRPC27309
Recombinant Canine parvovirus type 2 Capsid protein VP2 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. H ost: E. coli. Endotoxin Level: Not Tested. Species: Canine parvovirus type 2 (CPV-2). Target Name: Canine parvovirus 1 * 100 µG
  MSPPRPC27309
Frequently Bought Together