Recombinant Human Legumain (LGMN) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxi n Level: Not Tested. Species: Human (Homo sapiens). Target Name: LGMN. Target Synonyms: Asparaginyl endopeptidaseProt 1 * 100 µG
Catalog # MSPPRPC21601
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together