Human Glucoprotein 130 (gp130) ELISA Kit for detecting Glucoprotein 130 (gp130) in Serum, Plasma, Other Biological Fluids. Species R eactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKL61903
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together