Rat Catalase (CAT) ELISA Kit for detecting Catalase (CAT) in Serum, Plasma, Tissue Homogenates, Other Biological Fluids. Species Rea ctivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKL61793
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together