Human ESM1 (Endothelial Cell Specific Molecule 1) ELISA Kit for detecting ESM1 in serum, plasma, other biological fluids. Species Re activity: Human (Homo sapiens). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKE60936
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together