Order Entry
United Kingdom
ContactUsLinkComponent
Human DBP/Vitamin D Binding Protein (Vitamin D Binding Protein) ELISA Kit for detecting DBP Vitamin D Binding Protein in serum, pla sma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapien 1 * 96 Tests
  MSPPEKF57588
Human DBP/Vitamin D Binding Protein (Vitamin D Binding Protein) ELISA Kit for detecting DBP Vitamin D Binding Protein in serum, pla sma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapien 1 * 96 Tests
  MSPPEKF57588
Frequently Bought Together