Human CHCHD2 (Coiled-coil-helix-coiled-coil-helix domain-containing protein 2) ELISA Kit for detecting CHCHD2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detec 1 * 96 Tests
Catalog # MSPPEKF60802
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together