DA (Dopamine) ELISA Kit for detecting DA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKF57981
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together