Human Catenin Beta-1 (Ctnnb1/Ctnnb/Ok/Sw-Cl.35/Pro2286) Elisa Kit for detecting Catenin (Cadherin-Associated Protein), Beta 1, 88Kda in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens 1 * 96 Tests
Catalog # MSPPEKC40547
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together