Human Visfatin Elisa Kit for detecting Nicotinamide Phosphoribosyltransferase in serum, plasma, urine, tissue homogenates. Species R eactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKC40542
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together