Rat Cardiac Troponin I (Ctn-I) Elisa Kit for detecting Cardiac Troponin I(Ctn-I) in serum, plasma, tissue homogenates. Species React ivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKC40482
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together