Human Adenosine Deaminase (ADA) ELISA Kit for detecting ADA in serum, plasma, tissue homogenates, other biological fluids. Species R eactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKU02131
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together