Human Sequestosome 1 (SQSTM1) ELISA Kit for detecting SQSTM1 in tissue homogenates, cell lysates, other biological fluids. Species R eactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKU07279
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together