Mouse N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) Mini sample ELISA Kit for detecting NT-ProBNP in serum, plasma, tissue ho mogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). 1 * 96 Tests
Catalog # MSPPEKU11896
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together