Human/Mouse/Rat Slit Homolog 3 (Slit3) ELISA Kit for detecting Slit3 in serum, plasma, tissue homogenates, cell lysates, cell cultur e supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus no 1 * 96 Tests
Catalog # MSPPEKU09161
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together