Human Telomerase Reverse Transcriptase (TERT) ELISA Kit for detecting TERT in tissue homogenates, cell lysates, other biological flu ids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-We 1 * 96 Tests
Catalog # MSPPEKU07587
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together