Human Olfactomedin 4 (OLFM4) ELISA Kit for detecting OLFM4 in serum, plasma, tissue homogenates, other biological fluids. Species Re activity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.4pg/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKU06386
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together