Human/Mouse/Rat Angiotensin 1-7 (Ang1-7) ELISA Kit for detecting Ang1-7 in serum, plasma, tissue homogenates, cell lysates, cell cul ture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus 1 * 96 Tests
Catalog # MSPPEKU08763
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together