Order Entry
United Kingdom
ContactUsLinkComponent
Human Laminin Gamma 2 (LAMC2) ELISA Kit for detecting LAMC2 in serum, plasma, tissue homogenates, other biological fluids. Species R eactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKU05562
Human Laminin Gamma 2 (LAMC2) ELISA Kit for detecting LAMC2 in serum, plasma, tissue homogenates, other biological fluids. Species R eactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKU05562
Frequently Bought Together