Mouse Transferrin Receptor (TFR) ELISA Kit for detecting TFR in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assa 1 * 96 Tests
Catalog # MSPPEKU07789
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together