Human Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF) ELISA Kit for detecting GM-CSF in serum, plasma, tissue homogenat es, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detecti 1 * 96 Tests
Catalog # MSPPEKN44470
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together