Human Lectin Like Oxidized Low Density Lipoprotein Receptor 1 (LOX1) ELISA Kit for detecting LOX1 in serum, plasma, tissue homogenat es, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detecti 1 * 96 Tests
Catalog # MSPPEKN46687
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together