Order Entry
United Kingdom
ContactUsLinkComponent
Muffle Furnace L5/14/B510 Tmax1400 with flap door and protective gas connection and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3309
Supplier:  NABERTHERM
New Product
Muffle Furnace L5/14/B510 Tmax1400 with flap door and protective gas connection and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3309
Supplier:  NABERTHERM