Order Entry
United Kingdom
ContactUsLinkComponent
Muffle Furnace L15/13/B510 Tmax1300 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
Catalog # 455-3260
Supplier:  NABERTHERM
New Product
Muffle Furnace L15/13/B510 Tmax1300 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
Catalog # 455-3260
Supplier:  NABERTHERM